Assay Details
| Gene Symbol: Olfr221 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001808.2 | NP_001001808.2 | 1 | 59 | 71 | |
| GenBank mRNA | BC147427.1 | - | 1 | 93 | 9056495 | 71 |
| BC147428.1 | - | 1 | 93 | 9056478 | 71 | |
| BC151091.1 | - | 1 | 365 | 9088004 | 71 | |
| BC151095.1 | - | 1 | 358 | 9088008 | 71 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1953112 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P173168 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1953112 | - | - |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P109232 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29604418 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29604422 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29604414 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29604425 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29604413 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29604420 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29604421 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29604424 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29604426 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29604411 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P173168 | - | - |
| Whole Mouse Genome G4122A | A_52_P109232 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P133912 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P173168 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11683968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29604397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29604420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29604456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29604411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29604435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29604404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29604422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29604380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29604454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29604408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29604379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29604446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29604455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29604386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29604442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29604466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29604413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29604430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29604444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29604424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29604450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29604457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29604417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29604418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29604458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29604427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29604421 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29604422 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29604444 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29604447 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29604414 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29604458 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29604450 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29604448 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29604421 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29604420 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29604413 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29604401 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29604428 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29604440 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11732700 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29604417 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11207069 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11122907 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29604435 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29604398 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29604411 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29604433 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11829425 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29604424 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29604454 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29604407 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11126343 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29604410 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29604431 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29604404 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29604427 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29604457 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29604456 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11806343 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29604446 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11683962 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29604397 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29604405 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10419605 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10419604 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1400015146.mm.1 | TC1400002022.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1400015148.mm.1 | TC1400002022.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1400015147.mm.1 | NM_001001808.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400015147.mm.1 | NM_001001808.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400015147.mm.1 | TC1400002022.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1400015149.mm.1 | TC1400002022.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2664173 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2664173 | - | - |

Olfr221
Genomic Map