Assay Details
Gene Symbol: Olfr450 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146445.1 | NP_666656.1 | 1 | 457 | 116 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P468639 | NM_146445.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P468639 | NM_146445.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P168729 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P468639 | NM_146445.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P468639 | NM_146445.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P468639 | NM_146445.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P468639 | NM_146445.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24400279 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24400257 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24400277 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24400278 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24400272 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24400270 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24400258 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24400253 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24400256 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24400276 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24400265 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24400288 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24400293 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24400291 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24400282 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24400284 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24400251 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24400269 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24400290 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24400274 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24400280 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24400261 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24400246 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24400294 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24400247 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24400264 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24400285 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24400267 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24400287 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24400259 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24400268 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24400250 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24400266 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24400248 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24400254 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P168729 | - | - |
Whole Mouse Genome G4122A | A_52_P468639 | NM_146445.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P468639 | NM_146445.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P137770 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P168729 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05836931 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05836953 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05836942 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05836952 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05836949 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24400237 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24400280 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24400228 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24400264 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24400330 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24400236 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24400284 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24400282 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24400276 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24400269 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24400314 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24400327 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24400251 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24400235 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24400290 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24400333 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24400241 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24400231 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24400298 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24400265 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24400257 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24400285 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24400239 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24400253 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24400250 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24400288 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24400266 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24400309 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24400272 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24400246 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24400247 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24400297 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24400277 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24400306 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24400256 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24400311 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24400315 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24400270 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24400274 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24400259 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24400258 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24400313 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24400287 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24400267 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24400335 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24400268 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24400243 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24400279 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24400326 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24400310 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24400301 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24400255 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24400304 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24400322 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24400319 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24400242 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24400243 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24400290 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24400246 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24400288 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24400244 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24400233 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24400235 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24400270 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24400277 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24400272 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05836945 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05360453 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24400278 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24400228 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24400274 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05495481 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05495480 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05185907 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24400282 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24400276 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24400297 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24400313 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24400257 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24400294 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24400239 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24400247 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24400315 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24400306 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24400295 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24400262 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24400304 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24400284 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05185911 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24400301 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24400311 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24400287 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05207671 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24400269 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24400264 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24400231 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24400250 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24400259 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24400236 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24400300 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24400309 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24400251 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24400280 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24400310 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24400298 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24400266 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24400314 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10537800 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10537799 | - | - |
Affymetrix Mouse Clariom S Array | PSR0600003736.mm.1 | TC0600000468.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600003736.mm.1 | NM_146445.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600003736.mm.1 | NM_146445.1 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 525858 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 1770685 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 1770685 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0001770685 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1240945 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1240945 | - | - |
mouse-6 v1.1 (Illumina) | scl0258437.1_213-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 1770685 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0258437.1_213-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1770685 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1240945 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1240945 | - | - |
