Assay Details
| Gene Symbol: Olfr693 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146453.2 | NP_666664.1 | 1 | 902 | 93 | |
| GenBank mRNA | BC130248.1 | - | 1 | 905 | 40142368 | 93 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2146947 | BC130248.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2146947 | NM_146453.2 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P109038 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2146947 | BC130248.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2146947 | NM_146453.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P289976 | BC130248.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P289976 | NM_146453.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396272 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396277 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396283 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396284 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396281 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396258 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396261 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396290 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396286 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396318 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396257 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396264 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396293 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396254 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396278 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396262 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396323 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396294 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396273 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396288 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396265 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396256 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396298 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396275 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396259 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396320 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396292 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396280 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396282 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396324 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396266 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396287 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396325 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396279 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396296 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396321 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P109038 | - | - |
| Whole Mouse Genome G4122A | A_51_P289976 | BC130248.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P289976 | NM_146453.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P113232 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P109038 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06321832 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06597387 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06597396 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06597402 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06597391 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06597392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396288 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396309 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396264 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396261 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396256 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06364689 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396299 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396318 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396294 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396301 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396250 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396245 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396262 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396247 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396273 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396275 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396283 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06616374 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396311 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396282 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396281 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396243 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396249 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396266 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396313 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396292 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06597392 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396302 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396252 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396279 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396298 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396288 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396239 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396296 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396240 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396277 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396257 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396253 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396304 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396324 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396286 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396272 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396308 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396305 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396284 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396325 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06597388 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396317 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396246 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396258 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396293 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396254 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396307 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396290 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396241 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396323 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396315 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396320 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10566588 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10566587 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700033436.mm.1 | NM_146453.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700033436.mm.1 | NM_146453.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700033436.mm.1 | TC0700004013.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2650110 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002650110 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2739637 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2739637 | - | - |
| mouse-6 v1.1 (Illumina) | scl30865.1.1_21-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2650110 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl30865.1.1_21-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2650110 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2739637 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2739637 | - | - |

Olfr693
Genomic Map