Assay Details
| Gene Symbol: Olfr1196 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146464.2 | NP_666675.2 | 1 | 432 | 69 | |
| Gene Symbol: Olfr1197 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005225.1 | NP_001005225.1 | 1 | 432 | 69 | |
| Gene Symbol: Olfr1200 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005227.2 | NP_001005227.2 | 1 | 432 | 69 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2092291 | NM_146464.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2081525 | NM_001005227.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2133756 | NM_001005225.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2092291 | NM_146464.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2133756 | NM_001005225.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2081525 | NM_001005227.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P98537 | - | - |
| Whole Mouse Genome G4122A | A_52_P98537 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P138763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21410917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21410792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21410824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21410809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21410811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21410859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21410879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21410817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21410903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21410855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21410778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21410854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21410869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21410915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21410892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21410918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21410769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21410931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21410786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21410765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21410805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21410900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21410777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21410852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21410846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21410756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21410790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21410897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21410780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21410932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21410826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21410877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21410876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21410771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21410898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21410922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21410874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21410775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21410821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21410770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21410813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21410886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21410889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21410945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21410920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21410896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21410861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21410788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21410761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21410928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21410849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21410870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21410774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21410933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21410766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21410776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21410872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21410815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21410881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21410847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21410807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21411130 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410898 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410780 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410781 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410896 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01485068 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410811 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410775 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21411110 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410790 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410876 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410765 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410870 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410921 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410929 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21411130 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02170261 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01922190 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410792 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02180336 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410817 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410822 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410874 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410774 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410805 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21411125 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21411140 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410918 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410943 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410760 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410802 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410807 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410922 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410867 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01429155 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410783 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410788 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410892 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01346758 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21411115 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410894 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410938 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410776 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410942 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410756 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410877 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410824 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410891 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21411108 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410769 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21411142 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410900 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410926 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410761 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410766 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01325740 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410931 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410872 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21411139 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02053037 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410920 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410879 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410778 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21411124 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21411106 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21411136 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02180340 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410782 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21411129 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410777 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21411121 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01319468 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21411128 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01830227 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01963528 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21411137 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410886 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410915 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410928 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410935 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410827 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01319487 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410945 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02170257 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410869 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410770 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410786 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02040240 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410881 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410914 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410821 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410871 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410940 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410815 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410933 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410913 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21411127 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01591270 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410809 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410883 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410889 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21410813 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10484763 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10484757 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10484757 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10484755 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10484755 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10484754 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10484756 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10484762 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10484756 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10484754 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200033974.mm.1 | NM_001005227.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200033974.mm.1 | NM_001005227.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200033974.mm.1 | TC0200004028.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200033965.mm.1 | NM_146464.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200033965.mm.1 | NM_146464.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200033965.mm.1 | TC0200004024.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200033966.mm.1 | NM_001005225.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200033966.mm.1 | NM_001005225.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200033966.mm.1 | TC0200004025.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4850551 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4850551 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004850551 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1223930 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1223930 | - | - |
| mouse-6 v1.1 (Illumina) | scl0258456.1_19-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4850551 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0258456.1_19-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4850551 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2876794 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1223930 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2962810 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2876794 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1223930 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2962810 | - | - |

Olfr1196
Genomic Map