Assay Details
Gene Symbol: Olfr357 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146623.1 | NP_666834.1 | 1 | 851 | 76 | |
GenBank mRNA | BC119361.1 | - | 1 | 936 | 8734110 | 76 |
BC119363.1 | - | 1 | 936 | 8734112 | 76 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1976614 | NM_146623.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1976614 | BC119361.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1976614 | BC119363.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P106712 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1976614 | NM_146623.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1976614 | BC119361.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1976614 | BC119363.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P340525 | NM_146623.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P340525 | BC119361.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P340525 | BC119363.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140578 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140614 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140580 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140572 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140611 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140625 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140615 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140576 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140586 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140585 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140602 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140607 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140604 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140595 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140590 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140617 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140591 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140618 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140600 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140621 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140583 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140575 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140588 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140606 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140616 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140593 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140573 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140622 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140609 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140601 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140598 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140612 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140592 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140597 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140624 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P106712 | - | - |
Whole Mouse Genome G4122A | A_51_P340525 | NM_146623.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P340525 | BC119361.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P340525 | BC119363.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P128777 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P106712 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02018558 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02018568 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02018559 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02018565 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02018570 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140598 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140615 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140624 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140572 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140596 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140602 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140646 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140588 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140671 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140663 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140606 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140628 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140601 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140622 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140591 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140563 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140664 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140609 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140612 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140586 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140604 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140593 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140614 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140645 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140621 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140565 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140625 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140668 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140617 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140569 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140590 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140667 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140661 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140666 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140616 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140633 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140642 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140585 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140650 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140627 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140583 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140630 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140648 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140568 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140576 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140618 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140579 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140611 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140659 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140632 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140633 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140596 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140579 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140626 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140601 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140609 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140611 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140630 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01784931 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140566 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140569 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01291448 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140625 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140592 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140604 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140618 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140606 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140602 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140588 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140622 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140615 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140591 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140598 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01580764 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140614 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140570 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140600 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140616 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140624 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140617 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01572011 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140597 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01291450 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140565 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140567 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140595 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140612 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140590 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01784924 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140568 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140572 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140575 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140581 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140621 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01784920 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02275816 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10471771 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10471770 | - | - |
Affymetrix Mouse Clariom S Array | PSR0200006047.mm.1 | TC0200000676.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 372637 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 5360114 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0005360114 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2753127 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2753127 | - | - |
mouse-6 v1.1 (Illumina) | scl21007.1.1_30-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 5360114 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl21007.1.1_30-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5360114 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2753127 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2753127 | - | - |
