Assay Details
| Gene Symbol: Olfr152 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146646.2 | NP_666857.2 | 1 | 867 | 126 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2046268 | NM_146646.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2046268 | NM_146646.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P171458 | NM_146646.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21407182 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21407207 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21407199 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21407211 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21407202 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21407197 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21407189 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21407196 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21407216 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21407175 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21407186 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21407187 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21407180 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21407178 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21407212 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21407209 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21407213 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21407185 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21407179 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21407205 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21407214 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21407221 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21407200 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21407192 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21407194 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21407198 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21407206 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21407203 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21407183 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21407219 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21407176 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21407190 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04596965 | - | - |
| Whole Mouse Genome G4122A | A_52_P171458 | NM_146646.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129139 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P04596965 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01816371 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01816373 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01958971 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01816374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21407198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21407243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21407182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21407250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21407215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21407245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21407176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21407186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21407239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21407174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21407211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21407192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21407234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21407227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21407216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21407197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21407172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21407247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21407232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21407224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21407252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21407178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21407223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21407189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21407226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21407187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21407183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21407221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21407251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21407249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21407180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21407203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21407213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21407202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21407175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21407205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21407235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21407199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21407196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21407229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21407212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21407185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21407231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21407209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21407194 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21407192 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21407214 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21407232 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01678744 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21407226 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01875070 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21407175 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21407207 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21407176 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21407209 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21407185 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21407206 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21407243 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21407180 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21407251 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21407223 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21407215 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21407224 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21407211 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21407198 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21407221 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21407212 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21407186 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21407239 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21407193 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02088795 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21407174 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21407240 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21407213 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21407179 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21407202 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21407203 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21407172 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21407249 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21407182 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21407247 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21407194 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21407197 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21407219 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21407245 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21407205 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21407183 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21407178 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21407246 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21407250 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21407200 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21407235 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21407196 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21407229 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21407231 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21407234 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21407227 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21407199 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02003618 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21407189 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10473563 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10473562 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200010074.mm.1 | NM_146646.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200010074.mm.1 | NM_146646.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200010074.mm.1 | TC0200001301.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 100060195 | - | - |
| MouseWG-6 v2 Expression BeadChip | 380025 | AK147367.1 | genbank_mrna_mouse |
| MouseWG-6 v2 Expression BeadChip | 380025 | AK156295.1 | genbank_mrna_mouse |
| MouseWG-6 v2 Expression BeadChip | 380025 | AK161486.1 | genbank_mrna_mouse |
| MouseWG-6 v2 Expression BeadChip | 380025 | AK170640.1 | genbank_mrna_mouse |
| MouseWG-6 v2 Expression BeadChip | 380025 | AK171350.1 | genbank_mrna_mouse |
| MouseWG-6 v2 Expression BeadChip | 380025 | BC035536.1 | genbank_mrna_mouse |
| MouseWG-6 v2 Expression BeadChip | 380025 | NM_001033238.1 | refseq_mrna_mouse_NM |
| Illumina mouse-6 v1.0 expression beadchip | 380025 | AK147367.1 | genbank_mrna_mouse |
| Illumina mouse-6 v1.0 expression beadchip | 380025 | AK156295.1 | genbank_mrna_mouse |
| Illumina mouse-6 v1.0 expression beadchip | 380025 | AK161486.1 | genbank_mrna_mouse |
| Illumina mouse-6 v1.0 expression beadchip | 380025 | AK170640.1 | genbank_mrna_mouse |
| Illumina mouse-6 v1.0 expression beadchip | 380025 | AK171350.1 | genbank_mrna_mouse |
| Illumina mouse-6 v1.0 expression beadchip | 380025 | BC035536.1 | genbank_mrna_mouse |
| Illumina mouse-6 v1.0 expression beadchip | 380025 | NM_001033238.1 | refseq_mrna_mouse_NM |
| Illumina mouse-6 v1.0 expression beadchip | 100060195 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000380025 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1258236 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2775749 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1258236 | - | - |
| mouse-6 v1.1 (Illumina) | scl0258640.1_158-S | - | - |
| mouse-6 v1.1 (Illumina) | scl00258640.2_206-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 380025 | AK147367.1 | genbank_mrna_mouse |
| Illumina Sentrix mouse-6.v1 bead array | 380025 | AK156295.1 | genbank_mrna_mouse |
| Illumina Sentrix mouse-6.v1 bead array | 380025 | AK161486.1 | genbank_mrna_mouse |
| Illumina Sentrix mouse-6.v1 bead array | 380025 | AK170640.1 | genbank_mrna_mouse |
| Illumina Sentrix mouse-6.v1 bead array | 380025 | AK171350.1 | genbank_mrna_mouse |
| Illumina Sentrix mouse-6.v1 bead array | 380025 | BC035536.1 | genbank_mrna_mouse |
| Illumina Sentrix mouse-6.v1 bead array | 380025 | NM_001033238.1 | refseq_mrna_mouse_NM |
| Illumina Sentrix mouse-6.v1 bead array | 100060195 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00258640.2_206-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 380025 | AK147367.1 | genbank_mrna_mouse |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 380025 | AK156295.1 | genbank_mrna_mouse |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 380025 | AK161486.1 | genbank_mrna_mouse |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 380025 | AK170640.1 | genbank_mrna_mouse |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 380025 | AK171350.1 | genbank_mrna_mouse |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 380025 | BC035536.1 | genbank_mrna_mouse |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 380025 | NM_001033238.1 | refseq_mrna_mouse_NM |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 100060195 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1258236 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1258236 | - | - |

Olfr152
Genomic Map