Assay Details
| Gene Symbol: Akr1b10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172398.3 | NP_765986.3 | 9 - 10 | 1005 | 144 | |
| GenBank mRNA | AK019906.1 | - | 9 - 10 | 901 | 144 | |
| AK075865.1 | - | 9 - 10 | 1007 | 144 | ||
| AK082777.1 | - | 9 - 10 | 963 | 144 | ||
| AK087002.1 | - | 9 - 10 | 991 | 144 | ||
| BC037690.1 | - | 9 - 10 | 1012 | 3661180 | 144 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2095196 | BC037690.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2095196 | BY430637.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2095196 | AK075865.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2095196 | NM_172398.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2095196 | AK082777.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P486618 | BC037690.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P486618 | BY430637.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P486618 | AK019906.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P486618 | AK075865.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P486618 | NM_172398.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P486618 | BC037690.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P486618 | BY430637.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P486618 | AK019906.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P486618 | AK075865.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P486618 | NM_172398.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 837500 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 98492 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 382765 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 368881 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 122821 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 413021 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 951927 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 162167 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P158530 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P138166 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2095196 | BC037690.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2095196 | BY430637.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2095196 | AK075865.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2095196 | NM_172398.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2095196 | AK082777.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P486618 | BC037690.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P486618 | BY430637.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P486618 | AK019906.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P486618 | AK075865.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P486618 | NM_172398.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P486618 | BC037690.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P486618 | BY430637.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P486618 | AK019906.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P486618 | AK075865.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P486618 | NM_172398.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24355372 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24355399 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24355388 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24355384 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24355374 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24355377 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24355396 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24355389 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24355380 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24355366 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24355394 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24355385 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24355368 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24355376 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24355381 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24355390 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24355387 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24355397 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24355392 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P138166 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05859354 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P158530 | - | - |
| Whole Mouse Genome G4122A | A_52_P257058 | BC026843.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P486618 | BC037690.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P486618 | BY430637.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P486618 | AK019906.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P486618 | AK075865.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P486618 | NM_172398.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P486618 | BC037690.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P486618 | BY430637.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P486618 | AK019906.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P486618 | AK075865.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P486618 | NM_172398.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P120957 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P113981 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P138166 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P158530 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05348402 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05348405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24355449 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24355356 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24355399 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24355352 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24355384 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24355401 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24355403 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05109017 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24355409 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05395822 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24355390 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24355366 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24355402 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05659298 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24355372 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24355375 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05411188 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24355412 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24355416 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05457040 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24355423 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24355381 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24355376 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05361801 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05411193 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24355394 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24355408 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24355387 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24355350 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05411185 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24355380 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24355422 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24355397 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05448338 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24355363 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05361803 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24355428 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24355388 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05109018 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24355344 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05348403 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05669210 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05457039 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24355385 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05281805 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05348405 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24355347 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05659293 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05109010 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24355424 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05448339 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24355415 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10537160 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10537162 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10537163 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10537165 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10537167 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10537161 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10537158 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10537164 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10537168 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10537166 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10537159 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1425684_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1425685_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1453173_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10537157 | - | - |
| Mouse Genome 430 2.0 Array | 1425685_at | - | - |
| Mouse Genome 430 2.0 Array | 1425685_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1425684_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1425684_at | - | - |
| Mouse Genome 430 2.0 Array | 1453173_at | - | - |
| Mouse Genome 430 2.0 Array | 1453173_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67861_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67861_at | - | - |
| Mouse Expression Set 430 | 1425684_at | - | - |
| Mouse Expression Set 430 | 1425685_at | - | - |
| Mouse Expression Set 430 | 1425685_PM_at | - | - |
| Mouse Expression Set 430 | 1425684_PM_at | - | - |
| Mouse Expression Set 430 | 1453173_at | - | - |
| Mouse Expression Set 430 | 1453173_PM_at | - | - |
| Murine Genome U74v2 Set | 129503_at | - | - |
| Murine Genome U74v2 Set | 129503_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425685_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425685_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425684_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425684_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425684_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425685_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425685_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425684_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453173_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453173_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 785926 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3850113 | - | - |
| MouseWG-6 v2 Expression BeadChip | 2340068 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2340068 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3850113 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002340068 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003850113 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2733753 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2616422 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2733753 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2616422 | - | - |
| mouse-6 v1.1 (Illumina) | scl067861.1_19-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0067861.2_64-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3850113 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2340068 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0067861.2_64-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl067861.1_19-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2340068 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3850113 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2733753 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2616422 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2616422 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2733753 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0067861.2_64-S | - | - |

Akr1b10
Genomic Map