Assay Details
| Gene Symbol: Papola | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011112.3 | NP_035242.1 | 14 - 15 | 1494 | 86 | |
| GenBank mRNA | AK028940.1 | - | 14 - 15 | 1492 | 86 | |
| AK090096.1 | - | 14 - 15 | 1495 | 86 | ||
| AK132704.1 | - | 14 - 15 | 1481 | 86 | ||
| BC060128.1 | - | 13 - 14 | 1341 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Papola
Genomic Map