Assay Details
| Gene Symbol: Ecd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027475.3 | NP_081751.1 | 3 - 4 | 652 | 137 | |
| GenBank mRNA | AK028664.1 | - | 3 - 4 | 616 | 137 | |
| AK043261.1 | - | 3 - 4 | 501 | 137 | ||
| BC013470.1 | - | 3 - 4 | 601 | 4220504 | 137 | |
More Information
Set Membership: |
|

Ecd
Genomic Map