Assay Details
| Gene Symbol: Ecd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027475.3 | NP_081751.1 | 6 - 7 | 1120 | 106 | |
| GenBank mRNA | AK017681.1 | - | 4 - 5 | 544 | 106 | |
| AK028664.1 | - | 6 - 7 | 1084 | 106 | ||
| AK029169.1 | - | 5 - 6 | 2311 | 106 | ||
| AK043261.1 | - | 6 - 7 | 969 | 106 | ||
| BC013470.1 | - | 6 - 7 | 1069 | 4220504 | 106 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ecd
Genomic Map