Assay Details
| Gene Symbol: Ecd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027475.3 | NP_081751.1 | 7 - 8 | 1246 | 119 | |
| GenBank mRNA | AK017681.1 | - | 5 - 6 | 670 | 119 | |
| AK028664.1 | - | 7 - 8 | 1210 | 119 | ||
| AK043261.1 | - | 7 - 8 | 1095 | 119 | ||
| BC013470.1 | - | 7 - 8 | 1195 | 4220504 | 119 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ecd
Genomic Map