Assay Details
| Gene Symbol: Ecd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027475.3 | NP_081751.1 | 9 - 10 | 1462 | 121 | |
| GenBank mRNA | AK017681.1 | - | 7 - 8 | 886 | 121 | |
| AK028664.1 | - | 9 - 10 | 1426 | 121 | ||
| AK043261.1 | - | 9 - 10 | 1311 | 121 | ||
| BC013470.1 | - | 9 - 10 | 1411 | 4220504 | 121 | |
More Information
Set Membership: |
|

Ecd
Genomic Map