Assay Details
| Gene Symbol: Srsf10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010178.3 | NP_034308.1 | 4 - 5 | 599 | 149 | |
| GenBank mRNA | AB015895.1 | - | 4 - 5 | 507 | 149 | |
| AF042383.1 | - | 4 - 5 | 511 | 149 | ||
| AK150734.1 | - | 4 - 5 | 533 | 149 | ||
| AK151567.1 | - | 4 - 5 | 538 | 149 | ||
| AK168571.1 | - | 4 - 5 | 522 | 149 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Srsf10
Genomic Map