Assay Details
| Gene Symbol: Pogk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175170.4 | NP_780379.2 | 2 - 3 | 453 | 80 | |
| GenBank mRNA | AK031716.1 | - | 2 - 3 | 406 | 80 | |
| AK048067.1 | - | 2 - 3 | 412 | 80 | ||
| AK053898.1 | - | 2 - 3 | 430 | 80 | ||
| AK122520.1 | - | 1 - 2 | 391 | 80 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK048067.1,AK053898.1,NM_175170.4 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Pogk
Genomic Map