| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0900004010.mm.1 |
NR_015605.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900004010.mm.1 |
NR_015605.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900004010.mm.1 |
TC0900000569.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900004012.mm.1 |
NR_015605.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900004012.mm.1 |
NR_015605.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900004012.mm.1 |
TC0900000569.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900004009.mm.1 |
NR_015605.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900004009.mm.1 |
NR_015605.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900004009.mm.1 |
TC0900000569.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900004011.mm.1 |
NR_015605.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900004011.mm.1 |
NR_015605.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900004011.mm.1 |
TC0900000569.mm.1 |
MTA TC |