Assay Details
| Gene Symbol: 2900052N01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015605.1 | 3 - 4 | 1643 | 104 | ||
| GenBank mRNA | AK038437.1 | - | 4 - 5 | 1358 | 104 | |
| AK049847.1 | - | 3 - 4 | 1643 | 104 | ||
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P67212 | AK049847.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P67212 | AK140089.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P67212 | BC054400.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P67212 | NR_015605.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P67212 | AK013669.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P67212 | AK049847.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P67212 | AK140089.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P67212 | BC054400.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P67212 | NR_015605.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P67212 | AK013669.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 588109 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 278146 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 477321 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 282241 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 852496 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 238454 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 675866 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 334054 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 365119 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 25976 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P138759 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P116095 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P138759 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P116095 | - | - |
| Whole Mouse Genome G4122A | A_52_P317102 | AK049847.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P317102 | NR_015605.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P317102 | AK038437.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P67212 | AK049847.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P67212 | AK140089.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P67212 | BC054400.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P67212 | NR_015605.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P67212 | AK013669.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P67212 | AK049847.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P67212 | AK140089.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P67212 | BC054400.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P67212 | NR_015605.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P67212 | AK013669.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P138759 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P116095 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482451 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482517 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482554 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482498 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482493 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482588 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482607 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07674683 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482488 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482510 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482519 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482555 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482582 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07879096 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482577 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482512 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482505 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482460 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07674688 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482494 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482502 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482459 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482560 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482447 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482518 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08162245 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482568 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482462 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482508 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482583 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482539 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08035118 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482466 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482573 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482609 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482614 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07541224 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482468 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482619 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08157981 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482426 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482616 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482453 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482441 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482474 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482435 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482485 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08169861 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482627 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482538 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482490 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482479 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07926186 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482448 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07674679 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482499 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07510509 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482450 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08195987 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07941645 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482611 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482454 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482534 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482464 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482500 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482593 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482455 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482431 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482632 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482629 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482525 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482562 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08157992 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482504 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482444 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482428 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482532 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482438 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08169863 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07510514 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482524 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482620 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07486262 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482594 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482483 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482445 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482480 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482603 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482457 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482471 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482558 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08169859 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482578 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482631 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482625 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482516 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482533 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482528 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482530 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482433 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482624 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482476 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482574 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482586 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07486270 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482522 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482556 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482470 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07571866 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482623 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26482580 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08039391 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT MG-430 PM Array Plate | 1457495_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1436231_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1445673_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1445673_at | - | - |
| Mouse Genome 430 2.0 Array | 1457495_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1445673_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1457495_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900004022.mm.1 | NR_015605.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900004022.mm.1 | NR_015605.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900004022.mm.1 | TC0900000569.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900004019.mm.1 | NR_015605.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900004019.mm.1 | NR_015605.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900004019.mm.1 | TC0900000569.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900004021.mm.1 | NR_015605.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900004021.mm.1 | NR_015605.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900004021.mm.1 | TC0900000569.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1457495_at | - | - |
| Mouse Expression Set 430 | 1445673_at | - | - |
| Mouse Expression Set 430 | 1457495_PM_at | - | - |
| Mouse Expression Set 430 | 1445673_PM_at | - | - |
| Murine Genome U74v2 Set | 168021_at | - | - |
| Murine Genome U74v2 Set | 168021_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1436231_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1445673_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1457495_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1445673_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1457495_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1436231_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 530340 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5050035 | - | - |

2900052N01Rik
Genomic Map