Assay Details
| Gene Symbol: Cabyr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181731.3 | NP_859420.2 | 4 - 5 | 614 | 85 | |
| GenBank mRNA | AF359383.1 | - | 4 - 5 | 614 | 85 | |
| AK075751.1 | - | 4 - 5 | 620 | 85 | ||
| BC107271.1 | - | 4 - 5 | 593 | 40052614 | 85 | |
| BC107272.1 | - | 4 - 5 | 594 | 40052615 | 85 | |
More Information
Set Membership: |
|

Cabyr
Genomic Map