Assay Details
| Gene Symbol: Tor1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133673.3 | NP_598434.2 | 2 - 3 | 491 | 64 | |
| GenBank mRNA | AJ297743.1 | - | 2 - 3 | 521 | 64 | |
| AK017290.1 | - | 2 - 3 | 510 | 64 | ||
| AK054404.1 | - | 2 - 3 | 522 | 64 | ||
| AK149754.1 | - | 2 - 3 | 512 | 64 | ||
| AK159843.1 | - | 2 - 3 | 490 | 64 | ||
| BC018456.1 | - | 2 - 3 | 490 | 3582246 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tor1b
Genomic Map