Assay Details
| Gene Symbol: H6pd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173371.4 | NP_775547.2 | 1 - 2 | 278 | 67 | |
| GenBank mRNA | AK045199.1 | - | 1 - 2 | 279 | 67 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK045199.1, NM_173371.4 transcripts.
Set Membership: |
|

H6pd
Genomic Map