Assay Details
| Gene Symbol: H2-Ea-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010381.2 | NP_034511.2 | 2 - 3 | 633 | 101 | |
| GenBank mRNA | AK158317.1 | - | 2 - 3 | 573 | 101 | |
| AY303781.1 | - | 2 - 3 | 529 | 101 | ||
| AY303782.1 | - | 2 - 3 | 529 | 101 | ||
| AY303783.1 | - | 2 - 3 | 529 | 101 | ||
| BC106107.1 | - | 2 - 3 | 638 | 3491637 | 101 | |
| M12818.1 | - | 2 - 3 | 532 | 101 | ||
| U13648.1 | - | 2 - 3 | 633 | 101 | ||
| U13649.1 | - | 2 - 3 | 633 | 101 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

H2-Ea-ps
Genomic Map