Assay Details
| Gene Symbol: H2-Ea-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010381.2 | NP_034511.2 | 3 - 4 | 797 | 142 | |
| GenBank mRNA | BC106107.1 | - | 3 - 4 | 802 | 3491637 | 142 |
| U13648.1 | - | 3 - 4 | 797 | 142 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

H2-Ea-ps
Genomic Map