Assay Details
| Gene Symbol: Cabyr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027687.2 | NP_081963.1 | 4 - 5 | 1443 | 64 | |
| GenBank mRNA | AF359382.1 | - | 4 - 5 | 1443 | 64 | |
| AK132770.1 | - | 4 - 5 | 1452 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cabyr
Genomic Map