Assay Details
| Gene Symbol: Cdk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007659.3 | NP_031685.2 | 7 - 8 | 896 | 81 | |
| GenBank mRNA | AK030231.1 | - | 7 - 8 | 897 | 81 | |
| AK051859.1 | - | 7 - 8 | 923 | 81 | ||
| AK135516.1 | - | 7 - 8 | 900 | 81 | ||
| AK168054.1 | - | 7 - 8 | 897 | 81 | ||
| BC005614.1 | - | 6 - 7 | 789 | 81 | ||
| BC024396.1 | - | 7 - 8 | 874 | 4977366 | 81 | |
| M38724.1 | - | 6 - 7 | 817 | 81 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Cdk1
Genomic Map