Assay Details
| Gene Symbol: Pde6h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023898.4 | NP_076387.2 | 1 - 2 | 60 | 88 | |
| GenBank mRNA | AK020922.1 | - | 1 - 2 | 61 | 88 | |
| BC058192.1 | - | 1 - 2 | 50 | 88 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK020922.1, NM_023898.4 transcripts.
Set Membership: |
|

Pde6h
Genomic Map