Assay Details
| Gene Symbol: Apip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019735.4 | NP_062709.3 | 2 - 3 | 275 | 108 | |
| GenBank mRNA | AB028863.1 | - | 2 - 3 | 167 | 108 | |
| AK077705.1 | - | 2 - 3 | 276 | 108 | ||
| AK145450.1 | - | 2 - 3 | 277 | 108 | ||
| BC028434.1 | - | 2 - 3 | 218 | 1245515 | 108 | |
More Information
Updated Mapping Information
Set Membership: |
|

Apip
Genomic Map