Assay Details
| Gene Symbol: Cdc34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177613.2 | NP_808281.1 | 2 - 3 | 488 | 95 | |
| GenBank mRNA | BC039160.1 | - | 2 - 3 | 488 | 4024472 | 95 |
| BC094502.1 | - | 2 - 3 | 304 | 5034261 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
protein polyubiquitination
cell cycle
protein ubiquitination
proteasome-mediated ubiquitin-dependent protein catabolic process
positive regulation of neuron apoptotic process
negative regulation of cAMP-mediated signaling
protein K48-linked ubiquitination
positive regulation of inclusion body assembly
nucleotide binding
ubiquitin-protein transferase activity
ATP binding
ligase activity
ubiquitin protein ligase binding
ubiquitin protein ligase activity
ubiquitin conjugating enzyme activity
protein polyubiquitination
cell cycle
protein ubiquitination
proteasome-mediated ubiquitin-dependent protein catabolic process
positive regulation of neuron apoptotic process
negative regulation of cAMP-mediated signaling
protein K48-linked ubiquitination
positive regulation of inclusion body assembly
nucleotide binding
ubiquitin-protein transferase activity
ATP binding
ligase activity
ubiquitin protein ligase binding
ubiquitin protein ligase activity
ubiquitin conjugating enzyme activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27278227 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27278225 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27278230 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27278224 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27278229 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2027312 | BC094502.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2027312 | AK153982.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2027312 | BC039160.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2027312 | NM_177613.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 131254 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 623989 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 93353 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P118141 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2027312 | BC094502.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2027312 | AK153982.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2027312 | BC039160.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2027312 | NM_177613.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278234 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278258 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278217 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278263 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278235 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278245 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278230 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278267 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278208 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278251 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278224 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278272 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278223 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278253 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278222 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278220 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278255 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278215 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278247 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278209 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278211 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278205 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278227 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278237 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278265 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278275 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278252 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278273 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278241 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278248 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278246 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278266 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278239 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278261 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278269 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278243 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278262 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P118141 | - | - |
| Whole Mouse Genome G4122A | A_52_P661496 | - | - |
| Whole Mouse Genome G4122A | A_52_P594894 | BC094502.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P594894 | BC039160.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P594894 | NM_177613.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P283175 | AK136078.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P118141 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08628211 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08628191 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08628195 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08628181 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08628196 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08628192 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08628197 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27278229 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27278225 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27278230 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27278227 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27278224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278314 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278179 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08910257 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278211 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278235 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08642747 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278270 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278258 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278237 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28034569 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08474531 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278253 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08642740 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278207 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278248 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08628192 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278202 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08628191 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278203 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08628181 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278269 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278252 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28034561 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09330104 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278251 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278220 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278263 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278255 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28034564 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08910268 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278234 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28034567 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28034568 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278275 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28034559 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08885195 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09042034 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278272 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08628189 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08555957 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278224 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28034566 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278246 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28034570 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278264 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278206 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278261 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08910266 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278239 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28034560 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278243 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278222 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28034572 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278217 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278273 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278233 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08819975 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278215 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278209 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10364456 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10364459 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380416 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10364458 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10380415 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10364455 | - | - |
| Mouse Genome 430 2.0 Array | 1434879_at | - | - |
| Mouse Genome 430 2.0 Array | 1434879_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 216150_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 216150_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000005848.mm.1 | NM_177613.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000005848.mm.1 | NM_177613.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000005848.mm.1 | TC1000000810.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000005850.mm.1 | NM_177613.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000005850.mm.1 | NM_177613.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000005850.mm.1 | TC1000000810.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000005852.mm.1 | NM_177613.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000005852.mm.1 | NM_177613.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000005852.mm.1 | TC1000000810.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000005846.mm.1 | NM_177613.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000005846.mm.1 | NM_177613.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000005846.mm.1 | TC1000000810.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000005849.mm.1 | NM_177613.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000005849.mm.1 | NM_177613.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000005849.mm.1 | TC1000000810.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000005851.mm.1 | NM_177613.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000005851.mm.1 | NM_177613.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000005851.mm.1 | TC1000000810.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000005853.mm.1 | NM_177613.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000005853.mm.1 | NM_177613.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000005853.mm.1 | TC1000000810.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1434879_at | - | - |
| Mouse Expression Set 430 | 1434879_PM_at | - | - |
| Murine Genome U74v2 Set | 94048_PM_at | - | - |
| Murine Genome U74v2 Set | 162339_PM_r_at | - | - |
| Murine Genome U74v2 Set | 94048_at | - | - |
| Murine Genome U74v2 Set | 162339_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1434879_at | - | - |
| Mouse Genome 430A 2.0 Array | 1434879_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 897538 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4570451 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4570451 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1190152 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004570451 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001190152 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2763116 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2724386 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2763116 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2724386 | - | - |
| mouse-6 v1.1 (Illumina) | scl0216150.5_158-S | - | - |
| mouse-6 v1.1 (Illumina) | scl00216150.1_133-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1190152 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4570451 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0216150.5_158-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00216150.1_133-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1190152 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4570451 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2724386 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2763115 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2763115 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2724386 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0216150.5_158-S | - | - |

Cdc34
Genomic Map