Assay Details
| Gene Symbol: Lamtor2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031248.3 | NP_112538.1 | 3 - 4 | 383 | 147 | |
| GenBank mRNA | AJ277386.1 | - | 3 - 4 | 353 | 147 | |
| AK019006.1 | - | 3 - 4 | 378 | 147 | ||
| BC031816.1 | - | 3 - 4 | 383 | 4189054 | 147 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22314478 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22314475 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22314476 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22314480 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22314477 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22314479 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22314474 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2085932 | AK019006.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2085932 | BC031816.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2085932 | NM_031248.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 896990 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P111211 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2085932 | AK019006.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2085932 | BC031816.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2085932 | NM_031248.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22314470 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22314464 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22314478 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22314461 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22314457 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22314474 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22314463 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22314480 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22314467 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22314468 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22314459 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22314453 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22314469 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22314472 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22314455 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22314465 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22314476 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P111211 | - | - |
| Whole Mouse Genome G4122A | A_51_P125385 | AK019006.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P125385 | BC031816.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P125385 | NM_031248.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P125385 | AJ277386.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P118426 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P111211 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03114844 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03114833 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03114837 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03114840 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22314476 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22314479 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22314480 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22314478 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22314477 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22314475 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22314474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314474 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22314456 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03114839 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22314457 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03114837 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22314467 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22314470 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22314455 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22314476 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22314453 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03114841 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22314463 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22314464 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22314459 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02721194 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03061991 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10499422 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10499423 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10499421 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10499424 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422797_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1435729_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10499420 | - | - |
| Mouse Genome 430 2.0 Array | 1422797_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1435729_at | - | - |
| Mouse Genome 430 2.0 Array | 1422797_at | - | - |
| Mouse Genome 430 2.0 Array | 1435729_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 83409_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 83409_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300017648.mm.1 | NM_031248.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300017648.mm.1 | NM_031248.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300017648.mm.1 | TC0300002317.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300017647.mm.1 | NM_031248.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300017647.mm.1 | NM_031248.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300017647.mm.1 | TC0300002317.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300017644.mm.1 | NM_031248.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300017644.mm.1 | NM_031248.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300017644.mm.1 | TC0300002317.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300017646.mm.1 | NM_031248.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300017646.mm.1 | NM_031248.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300017646.mm.1 | TC0300002317.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300017640.mm.1 | NM_031248.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300017640.mm.1 | NM_031248.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300017640.mm.1 | TC0300002317.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300017639.mm.1 | NM_031248.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300017639.mm.1 | NM_031248.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300017639.mm.1 | TC0300002317.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1422797_PM_at | - | - |
| Mouse Expression Set 430 | 1422797_at | - | - |
| Mouse Expression Set 430 | 1435729_PM_at | - | - |
| Mouse Expression Set 430 | 1435729_at | - | - |
| Murine Genome U74v2 Set | 99164_PM_at | - | - |
| Murine Genome U74v2 Set | 99164_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422797_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1435729_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422797_at | - | - |
| Mouse Genome 430A 2.0 Array | 1435729_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422797_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1435729_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422797_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1435729_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4480484 | - | - |
| MouseWG-6 v2 Expression BeadChip | 102940014 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4480484 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 102940019 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 102940014 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004480484 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1244381 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1225823 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1244381 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1236491 | - | - |
| mouse-6 v1.1 (Illumina) | scl21970.4.1_6-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4480484 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 102940019 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 102940014 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl21970.4.1_6-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4480484 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102940019 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102940014 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1244381 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2544661 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1244381 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2549544 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl21970.4.1_6-S | - | - |

Lamtor2
Genomic Map