Assay Details
| Gene Symbol: Timm21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025969.4 | NP_080245.1 | 1 - 2 | 528 | 165 | |
| GenBank mRNA | AK012203.1 | - | 1 - 2 | 528 | 165 | |
| AK017477.1 | - | 1 - 2 | 496 | 165 | ||
| AK032468.1 | - | 1 - 2 | 529 | 165 | ||
| AK168105.1 | - | 1 - 2 | 439 | 165 | ||
| BC034297.1 | - | 1 - 2 | 503 | 4511762 | 165 | |
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31891630 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31891633 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31891632 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P395842 | BC034297.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P395842 | AK029037.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P395842 | AK032468.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P395842 | AK168105.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P395842 | NM_025969.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P395842 | BC034297.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P395842 | AK029037.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P395842 | AK032468.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P395842 | AK168105.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P395842 | NM_025969.4 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P130822 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P395842 | BC034297.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P395842 | AK029037.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P395842 | AK032468.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P395842 | AK168105.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P395842 | NM_025969.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P395842 | BC034297.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P395842 | AK029037.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P395842 | AK032468.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P395842 | AK168105.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P395842 | NM_025969.4 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31891656 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31891663 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31891655 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31891636 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31891651 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31891615 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31891622 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31891638 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31891627 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31891610 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31891649 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31891662 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31891634 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31891658 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31891659 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31891642 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31891617 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31891629 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31891647 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31891640 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31891661 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31891631 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31891613 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31891632 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31891643 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31891611 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31891612 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31891606 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31891620 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31891625 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31891633 | - | - |
| Whole Mouse Genome G4122A | A_51_P395842 | BC034297.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P395842 | AK029037.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P395842 | AK032468.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P395842 | AK168105.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P395842 | NM_025969.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P395842 | BC034297.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P395842 | AK029037.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P395842 | AK032468.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P395842 | AK168105.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P395842 | NM_025969.4 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P102905 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13918711 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13918730 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13918721 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13918712 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13918731 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13918728 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13918727 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31891630 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31891632 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31891633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31891620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31891615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31891661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31891655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31891647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31891560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31891669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31891626 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31891678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31891606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31891656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31891629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31891677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31891550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31891601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31891597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31891691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31891700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31891613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31891622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31891627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31891697 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31891530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31891599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31891640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31891636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31891663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31891632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31891680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31891659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31891596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31891604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31891675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31891634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31891557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31891685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31891533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31891687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31891695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31891658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31891671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31891667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31891605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31891679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31891642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31891541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31891612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31891536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31891545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31891540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31891598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31891631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31891638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31891553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31891689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31891555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31891610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31891649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31891547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31891682 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31891607 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13765931 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31891618 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31891604 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13825436 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13614944 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31891663 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31891665 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31891622 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13918730 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31891661 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31891662 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31891615 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31891671 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31891651 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31891620 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13614950 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31891610 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31891645 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31891655 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31891605 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31891596 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13659042 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31891638 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31891643 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31891675 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31891649 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31891678 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31891597 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31891667 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31891679 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31891611 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31891601 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31891613 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31891633 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13918728 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31891626 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31891636 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13614945 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31891642 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14018975 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31891677 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31891656 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31891599 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31891617 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31891640 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14018986 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31891647 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13918721 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31891598 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31891634 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10608682 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460103 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460105 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460104 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1452785_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10608682 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10460102 | - | - |
| Mouse Genome 430 2.0 Array | 1452785_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1452785_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67105_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67105_PM_at | - | - |
| Mouse Expression Set 430 | 1452785_PM_at | - | - |
| Mouse Expression Set 430 | 1452785_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1452785_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1452785_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 333497 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 840021 | - | - |
| MouseWG-6 v2 Expression BeadChip | 6860026 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6860026 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6100348 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 840021 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000840021 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006100348 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006860026 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2685133 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2684388 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2732877 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2684388 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2685133 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2732877 | - | - |
| mouse-6 v1.1 (Illumina) | scl067105.1_68-S | - | - |
| mouse-6 v1.1 (Illumina) | scl067105.6_6-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0067105.1_17-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6860026 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 840021 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6100348 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl067105.6_6-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0067105.1_17-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl067105.1_68-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6100348 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 840021 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6860026 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2685133 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2732877 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2897476 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2684388 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2684388 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2685133 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2732877 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2897476 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl067105.6_6-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl067105.1_68-S | - | - |

Timm21
Genomic Map