Assay Details
| Gene Symbol: 4933434E20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027500.4 | NP_081776.2 | 5 - 6 | 765 | 116 | |
| GenBank mRNA | AK017833.1 | - | 5 - 6 | 733 | 116 | |
| BG091444.1 | - | 3 - 4 | 360 | 116 | ||
More Information
Updated Mapping Information
Set Membership: |
|

4933434E20Rik
Genomic Map