Assay Details
| Gene Symbol: 9330175E14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK034303.1 | - | 4248 | 116 | ||
| BC103768.1 | - | 2958 | 116 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2276224 | BC103768.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2276224 | AK034303.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 621366 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P132712 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2276224 | BC103768.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2276224 | AK034303.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P311976 | NR_015514.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P311976 | AK034303.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P311976 | AK053303.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P417945 | BC103768.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P417945 | AK034303.1 | mouse_transcriptome |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26077454 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26077458 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06916303 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07388952 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26077468 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26077460 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07427840 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26077461 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26077464 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07388950 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26077489 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26077484 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26077496 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26077453 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07405446 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26077501 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07388951 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26077498 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07405448 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26077500 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07365298 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26077473 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26077490 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26077495 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10580755 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1446547_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1460134_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10580754 | - | - |
| Mouse Genome 430 2.0 Array | 1460134_at | - | - |
| Mouse Genome 430 2.0 Array | 1446547_at | - | - |
| Mouse Genome 430 2.0 Array | 1460134_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1446547_PM_at | - | - |
| Mouse Expression Set 430 | 1460134_PM_at | - | - |
| Mouse Expression Set 430 | 1460134_at | - | - |
| Mouse Expression Set 430 | 1446547_at | - | - |
| Mouse Expression Set 430 | 1446547_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1446547_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1460134_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1446547_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1460134_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 373614 | - | - |

9330175E14Rik
Genomic Map