Assay Details
| Gene Symbol: Rnf141 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK011839.1 | - | 6 | 1871 | 114 | |
| AK139910.1 | - | 6 | 1880 | 114 | ||
| BC018553.1 | - | 6 | 1876 | 3582546 | 114 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rnf141
Genomic Map