Assay Details
| Gene Symbol: 2810417H13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026515.2 | NP_080791.2 | 4 | 1977 | 157 | |
| GenBank mRNA | AK013105.1 | - | 4 | 1937 | 157 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P133404 | AK013105.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P133404 | NM_026515.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2017870 | BC021413.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2017870 | BC096393.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2017870 | AK011835.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2017870 | AK012140.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2017870 | AK013105.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P157302 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P154277 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P133404 | AK013105.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P133404 | NM_026515.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2017870 | BC021413.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2017870 | BC096393.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2017870 | AK011835.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2017870 | AK012140.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2017870 | AK013105.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26590072 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26590082 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26590091 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26590083 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26590075 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26590093 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26590102 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26590088 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26590098 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26590086 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26590096 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26590080 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26590108 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26590085 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26590109 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26590105 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26590101 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26590097 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26590094 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26590076 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26590099 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26590078 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26590090 | - | - |
| Whole Mouse Genome G4122A | A_52_P13448 | BC021413.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P13448 | BC096393.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P13448 | AK011835.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P13448 | AK012140.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P13448 | AK013105.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P133404 | AK013105.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P133404 | NM_026515.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100916 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P104239 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08093982 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07641818 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08093966 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08061090 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08093981 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08061093 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08093977 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08061097 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07641815 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08061091 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08093970 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08093962 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08061101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590094 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26590211 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26590054 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07456262 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26590117 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26590063 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26590124 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07975659 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26590080 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26590093 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26590127 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08178369 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26590129 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07761956 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26590121 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26590055 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26590099 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08093969 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26590122 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26590075 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26590113 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07527470 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26590094 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26590078 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08061089 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26590070 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07774421 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26590058 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08137914 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26590066 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26590102 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26590110 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26590115 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26590109 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08061095 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26590059 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26590114 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26590118 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26590090 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26590088 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26590060 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26590108 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26590072 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08093966 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26590091 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26590083 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26590130 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26590064 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26590067 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26590112 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26590053 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08061093 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10586452 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10586453 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10586449 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10586450 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10350839 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10586451 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1419153_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1419152_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1431273_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10586448 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10350838 | - | - |
| Mouse Genome 430 2.0 Array | 1419153_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1419153_at | - | - |
| Mouse Genome 430 2.0 Array | 1431273_at | - | - |
| Mouse Genome 430 2.0 Array | 1431273_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1419152_at | - | - |
| Mouse Genome 430 2.0 Array | 1419152_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68026_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68026_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900006560.mm.1 | NM_026515.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900006560.mm.1 | NM_026515.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900006560.mm.1 | TC0900000854.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1419153_at | - | - |
| Mouse Expression Set 430 | 1419152_at | - | - |
| Mouse Expression Set 430 | 1419153_PM_at | - | - |
| Mouse Expression Set 430 | 1419152_PM_at | - | - |
| Mouse Expression Set 430 | 1431273_PM_at | - | - |
| Mouse Expression Set 430 | 1431273_at | - | - |
| Murine Genome U74v2 Set | 100116_PM_at | - | - |
| Murine Genome U74v2 Set | 100116_at | - | - |
| Murine Genome U74v2 Set | 161974_PM_r_at | - | - |
| Murine Genome U74v2 Set | 161974_r_at | - | - |
| Murine Genome U74v2 Set | 165853_i_at | - | - |
| Murine Genome U74v2 Set | 165853_PM_i_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419152_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419152_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419153_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419153_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419152_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419153_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419153_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419152_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431273_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431273_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 793711 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4850091 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4850091 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004850091 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1223045 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1223045 | - | - |
| mouse-6 v1.1 (Illumina) | scl36799.4_9-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4850091 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl36799.4_9-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4850091 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1223045 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1223045 | - | - |

2810417H13Rik
Genomic Map