Assay Details
| Gene Symbol: Kctd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK017384.1 | - | 3 | 1404 | 135 | |
| AK082924.1 | - | 3 | 1406 | 135 | ||
| AK159067.1 | - | 3 | 2818 | 135 | ||
| BC096370.1 | - | 3 | 1246 | 3597940 | 135 | |
| CJ248971.1 | - | 1 | 206 | 135 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29388379 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29388384 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29388383 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29388380 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29388381 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P297865 | AK082924.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P297865 | AK159067.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P297865 | BC096370.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P297865 | NM_027782.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P297865 | AK082924.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P297865 | AK159067.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P297865 | BC096370.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P297865 | NM_027782.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 159869 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 671311 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 946755 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 331907 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 372783 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 650024 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 39895 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P135799 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P297865 | AK082924.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P297865 | AK159067.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P297865 | BC096370.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P297865 | NM_027782.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P297865 | AK082924.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P297865 | AK159067.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P297865 | BC096370.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P297865 | NM_027782.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P297865 | AK082924.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P297865 | AK159067.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P297865 | BC096370.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P297865 | NM_027782.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P297865 | AK082924.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P297865 | AK159067.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P297865 | BC096370.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P297865 | NM_027782.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29388384 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29388361 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29388362 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29388360 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29388382 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29388371 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29388397 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29388403 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29388379 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29388367 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29388358 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29388377 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29388393 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29388398 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29388386 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29388355 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29388402 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29388391 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29388357 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29388380 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29388395 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29388390 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29388374 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29388375 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29388354 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29388364 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29388348 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29388401 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29388399 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29388352 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29388387 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29388347 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29388370 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29388389 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29388373 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29388366 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29388350 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P135799 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08021942 | - | - |
| Whole Mouse Genome G4122A | A_51_P297865 | AK082924.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P297865 | AK159067.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P297865 | BC096370.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P297865 | NM_027782.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P297865 | AK082924.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P297865 | AK159067.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P297865 | BC096370.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P297865 | NM_027782.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100681 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P135799 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11669205 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11373895 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29388380 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29388381 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29388383 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29388384 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29388379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29388389 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11434396 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29388402 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29388353 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29388372 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29388427 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29388415 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29388328 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29388386 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29388343 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11164068 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29388436 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29388366 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29388370 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29388387 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29388374 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29388441 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29388367 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29388432 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29388416 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29388393 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29388334 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29388337 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29388352 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29388424 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29388440 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29388330 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11147045 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29388412 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29388397 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29388344 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29388405 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11379065 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29388333 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11268119 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29388345 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29388361 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29388364 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29388384 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11213378 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29388395 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29388431 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29388360 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29388331 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29388409 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29388408 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11434397 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29388390 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29388406 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11164073 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11193979 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29388355 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29388335 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11246882 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29388403 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29388429 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29388430 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29388428 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29388341 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29388401 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29388433 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29388362 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29388377 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29388398 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29388438 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29388411 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29388425 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29388419 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29388389 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29388399 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11213382 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29388357 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10412654 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10412653 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10412651 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10412652 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428186_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10412650 | - | - |
| Mouse Genome 430 2.0 Array | 1428186_at | - | - |
| Mouse Genome 430 2.0 Array | 1428186_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71393_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71393_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1400000721.mm.1 | NM_027782.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400000721.mm.1 | TC1400000078.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1428186_at | - | - |
| Mouse Expression Set 430 | 1428186_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428186_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428186_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 497843 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4070435 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004070435 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2716236 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2716236 | - | - |
| mouse-6 v1.1 (Illumina) | scl46638.4_82-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4070435 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl46638.4_82-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4070435 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1247335 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2974117 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1247335 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2974117 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl46638.4_82-S | - | - |

Kctd6
Genomic Map