Assay Details
| Gene Symbol: Npm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008723.1 | NP_032749.1 | 6 | 727 | 158 | |
| GenBank mRNA | AK011749.1 | - | 6 | 730 | 158 | |
| AK146603.1 | - | 6 | 731 | 159 | ||
| AK157198.1 | - | 6 | 751 | 159 | ||
| U64450.1 | - | 6 | 727 | 158 | ||
| Gene Symbol: Npm3-ps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002702.1 | 1 | 771 | 171 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32140281 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32140280 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32140283 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32140284 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2075180 | NR_002702.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2010758 | AK157198.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2010758 | U64450.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2010758 | NM_008723.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2010758 | AK011749.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2010758 | AK146603.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P115140 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2010758 | AK157198.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2010758 | U64450.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2010758 | NM_008723.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2010758 | AK011749.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2010758 | AK146603.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P499201 | NR_002702.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P499201 | CK386398.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P499201 | AK157198.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P499201 | AK171848.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P499201 | BC048491.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32140303 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32140274 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32140284 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32140283 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32140298 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32140289 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32140288 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32140281 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32140297 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32140300 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32140302 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32140305 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32140285 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32140291 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32140304 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32140295 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32140301 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32140276 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32140277 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P115140 | - | - |
| Whole Mouse Genome G4122A | A_51_P499201 | NR_002702.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P499201 | CK386398.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P499201 | AK157198.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P499201 | AK171848.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P499201 | BC048491.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P115140 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14319236 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14439778 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14439762 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14439779 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14439766 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32140283 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32140280 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32140281 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32140284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32140274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32140240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32140238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32140194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32140220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32140291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32140190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32140234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32140200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32140191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32140203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32140219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32140284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32140202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32140289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32140228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32140277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32140205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32140213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32140248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32140244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32140206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32140283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32140245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32140250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32140276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32140281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32140239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32140231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32140197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32140196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32140233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32140215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32140246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32140212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32140249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32140222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32140242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32140285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32140218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32140288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32140211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32140223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32140207 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32140291 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32140276 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32140289 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32140288 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32140280 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32140285 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14319229 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32140277 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32140274 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10468087 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10468085 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10468088 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10468082 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539575 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10468086 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10468083 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10468087 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10468083 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539575 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10468084 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10468086 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10468084 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10468082 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10468088 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10468085 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10539574 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10468081 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10468081 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10539574 | - | - |
| Mouse Genome 430 2.0 Array | 1423522_at | - | - |
| Mouse Genome 430 2.0 Array | 1423522_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 108176_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 108176_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600007375.mm.1 | TC0600000954.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600007375.mm.1 | NR_002702.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900014055.mm.1 | NM_008723.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900014055.mm.1 | NM_008723.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900014055.mm.1 | TC1900001587.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900014061.mm.1 | NM_008723.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900014061.mm.1 | NM_008723.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900014061.mm.1 | TC1900001587.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900014058.mm.1 | NM_008723.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900014058.mm.1 | NM_008723.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900014058.mm.1 | TC1900001587.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900014057.mm.1 | NM_008723.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900014057.mm.1 | NM_008723.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900014057.mm.1 | TC1900001587.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900014054.mm.1 | NM_008723.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900014054.mm.1 | NM_008723.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900014054.mm.1 | TC1900001587.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900014053.mm.1 | NM_008723.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900014053.mm.1 | NM_008723.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900014053.mm.1 | TC1900001587.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900014059.mm.1 | NM_008723.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900014059.mm.1 | NM_008723.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900014059.mm.1 | TC1900001587.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1423522_at | - | - |
| Mouse Expression Set 430 | 1423522_at | - | - |
| Mouse Expression Set 430 | 1423522_PM_at | - | - |
| Mouse Expression Set 430 | 1423522_PM_at | - | - |
| Murine Genome U74v2 Set | 102796_at | - | - |
| Murine Genome U74v2 Set | 102796_PM_at | - | - |
| Murine Genome U74v2 Set | 102796_PM_at | - | - |
| Murine Genome U74v2 Set | 102796_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423522_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423522_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423522_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423522_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 896326 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3610300 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3610300 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003610300 | - | - |
| mouse-6 v1.1 (Illumina) | scl29824.1.5_102-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3610300 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl29824.1.5_102-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3610300 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3093150 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2965417 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3020829 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3093150 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3020829 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2688681 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2965417 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl29824.1.5_102-S | - | - |

Npm3
Genomic Map