Assay Details
| Gene Symbol: Med29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026042.3 | NP_080318.2 | 4 | 1065 | 144 | |
| GenBank mRNA | BC023192.1 | - | 4 | 1052 | 4947722 | 144 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25021604 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25021603 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25021601 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25021605 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25021607 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2036894 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P17492 | NM_026042.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P17492 | AK005112.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P17492 | AK013006.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P17492 | BC023192.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P17492 | BC083314.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 711333 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 331536 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 651607 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 132903 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 807921 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 77476 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2036894 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P17492 | NM_026042.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P17492 | AK005112.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P17492 | AK013006.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P17492 | BC023192.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P17492 | BC083314.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25021587 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25021596 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25021590 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25021589 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25021591 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25021606 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25021598 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25021595 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25021586 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25021603 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25021592 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25021581 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25021594 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25021579 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25021583 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25021599 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25021584 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06149378 | - | - |
| Whole Mouse Genome G4122A | A_51_P384584 | NM_026042.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P384584 | AK013006.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P384584 | BC023192.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P108797 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06149378 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06250450 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06250455 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25021607 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25021604 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25021605 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25021601 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25021603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25021535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25021606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25021509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25021589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25021531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25021501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25021567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25021547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25021519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25021530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25021586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25021560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25021537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25021511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25021591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25021549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25021529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25021584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25021516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25021594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25021521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25021514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25021600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25021494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25021525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25021603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25021596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25021569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25021571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25021581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25021496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25021540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25021528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25021492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25021497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25021550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25021506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25021579 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25021592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25021583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25021553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25021561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25021499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25021558 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05898920 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25021577 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25021589 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05914291 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25021594 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25021579 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25021567 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25021583 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25021588 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25021571 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25021587 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25021586 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25021569 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25021598 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05914281 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06168241 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25021581 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25021595 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06250455 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25021596 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25021572 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25021600 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10561457 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561458 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561460 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561459 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1421902_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1421903_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1436415_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10561456 | - | - |
| Mouse Genome 430 2.0 Array | 1421902_at | - | - |
| Mouse Genome 430 2.0 Array | 1421903_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1421902_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1421903_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67224_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67224_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700023616.mm.1 | TC0700002697.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700023616.mm.1 | NM_026042.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700023616.mm.1 | NM_026042.3 | Refseq |
| Mouse Expression Set 430 | 1421903_PM_at | - | - |
| Mouse Expression Set 430 | 1421903_at | - | - |
| Mouse Expression Set 430 | 1421902_PM_at | - | - |
| Mouse Expression Set 430 | 1421902_at | - | - |
| Mouse Expression Set 430 | 1436415_PM_at | - | - |
| Mouse Expression Set 430 | 1436415_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421903_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421902_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421902_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421903_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421902_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421903_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421903_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421902_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 739787 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2630100 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2630100 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002630100 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2615538 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2615538 | - | - |
| mouse-6 v1.1 (Illumina) | scl31581.5.1_37-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2630100 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl31581.5.1_37-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2630100 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2615538 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2615538 | - | - |

Med29
Genomic Map