Assay Details
| Gene Symbol: Calb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007586.1 | NP_031612.1 | 1 - 2 | 163 | 104 | |
| GenBank mRNA | AK032171.1 | - | 169 | 104 | ||
| BC017646.1 | - | 1 - 2 | 163 | 4527074 | 104 | |
More Information
Set Membership: |
|

Calb2
Genomic Map