Assay Details
| Gene Symbol: Cr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007758.2 | NP_031784.1 | 2 - 3 | 554 | 107 | |
| GenBank mRNA | AK003072.2 | - | 2 - 3 | 554 | 107 | |
| BC109344.1 | - | 2 - 3 | 487 | 40054528 | 107 | |
| M35684.1 | - | 1 - 2 | 422 | 107 | ||
| M61132.1 | - | 1 - 2 | 389 | 107 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cr2
Genomic Map