Assay Details
| Gene Symbol: Crabp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007759.2 | NP_031785.1 | 2 - 3 | 403 | 73 | |
| GenBank mRNA | BC018397.1 | - | 2 - 3 | 361 | 3588770 | 73 |
| M35523.1 | - | 2 - 3 | 370 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Crabp2
Genomic Map