Assay Details
| Gene Symbol: Emp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007929.2 | NP_031955.2 | 2 - 3 | 326 | 69 | |
| GenBank mRNA | AF083876.1 | - | 2 - 3 | 193 | 69 | |
| AF346627.1 | - | 2 - 3 | 268 | 69 | ||
| AK076194.1 | - | 2 - 3 | 305 | 69 | ||
| AK077260.1 | - | 2 - 3 | 328 | 69 | ||
| BC031646.1 | - | 1 - 2 | 119 | 69 | ||
| BC041670.1 | - | 2 - 3 | 229 | 3663158 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Emp2
Genomic Map