Assay Details
| Gene Symbol: Mcm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008563.2 | NP_032589.1 | 15 - 16 | 2155 | 62 | |
| GenBank mRNA | AK088142.1 | - | 14 - 15 | 2661 | 62 | |
| AK133850.1 | - | 15 - 16 | 2220 | 62 | ||
| AK145563.1 | - | 15 - 16 | 2254 | 62 | ||
| AK146497.1 | - | 15 - 16 | 2168 | 62 | ||
| AK147065.1 | - | 15 - 16 | 2170 | 62 | ||
| AK148006.1 | - | 15 - 16 | 2200 | 62 | ||
| AK192246.1 | - | 1 - 2 | 86 | 62 | ||
| AK196855.1 | - | 1 - 2 | 86 | 62 | ||
| AK216175.1 | - | 1 - 2 | 86 | 62 | ||
| BC020400.1 | - | 15 - 16 | 2170 | 62 | ||
| BC031700.1 | - | 15 - 16 | 2155 | 5007886 | 62 | |
| X62154.1 | - | 14 - 15 | 2099 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mcm3
Genomic Map