Assay Details
| Gene Symbol: Mcm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008563.2 | NP_032589.1 | 1 - 2 | 86 | 78 | |
| GenBank mRNA | AK088142.1 | - | 1 - 2 | 135 | 78 | |
| AK145563.1 | - | 1 - 2 | 185 | 78 | ||
| AK146497.1 | - | 1 - 2 | 99 | 78 | ||
| AK147065.1 | - | 1 - 2 | 101 | 78 | ||
| AK148006.1 | - | 1 - 2 | 131 | 78 | ||
| AK190847.1 | - | 1 - 2 | 45 | 78 | ||
| BC020400.1 | - | 1 - 2 | 101 | 78 | ||
| BC031700.1 | - | 1 - 2 | 86 | 5007886 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mcm3
Genomic Map