Assay Details
| Gene Symbol: Mcm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008563.2 | NP_032589.1 | 4 - 5 | 540 | 83 | |
| GenBank mRNA | AK088142.1 | - | 4 - 5 | 589 | 83 | |
| AK133850.1 | - | 4 - 5 | 605 | 83 | ||
| AK145563.1 | - | 4 - 5 | 639 | 83 | ||
| AK146497.1 | - | 4 - 5 | 553 | 83 | ||
| AK147065.1 | - | 4 - 5 | 555 | 83 | ||
| AK148006.1 | - | 4 - 5 | 585 | 83 | ||
| BC020400.1 | - | 4 - 5 | 555 | 83 | ||
| BC031700.1 | - | 4 - 5 | 540 | 5007886 | 83 | |
| X62154.1 | - | 3 - 4 | 484 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mcm3
Genomic Map