Assay Details
| Gene Symbol: Mcm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008563.2 | NP_032589.1 | 8 - 9 | 1163 | 80 | |
| GenBank mRNA | AK088142.1 | - | 8 - 9 | 1212 | 80 | |
| AK133850.1 | - | 8 - 9 | 1228 | 80 | ||
| AK145563.1 | - | 8 - 9 | 1262 | 80 | ||
| AK146497.1 | - | 8 - 9 | 1176 | 80 | ||
| AK147065.1 | - | 8 - 9 | 1178 | 80 | ||
| AK148006.1 | - | 8 - 9 | 1208 | 80 | ||
| BC020400.1 | - | 8 - 9 | 1178 | 80 | ||
| BC031700.1 | - | 8 - 9 | 1163 | 5007886 | 80 | |
| X62154.1 | - | 7 - 8 | 1107 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mcm3
Genomic Map