Assay Details
| Gene Symbol: Mcm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008563.2 | NP_032589.1 | 9 - 10 | 1377 | 85 | |
| GenBank mRNA | AK088142.1 | - | 9 - 10 | 1426 | 85 | |
| AK133850.1 | - | 9 - 10 | 1442 | 85 | ||
| AK145563.1 | - | 9 - 10 | 1476 | 85 | ||
| AK146497.1 | - | 9 - 10 | 1390 | 85 | ||
| AK147065.1 | - | 9 - 10 | 1392 | 85 | ||
| AK148006.1 | - | 9 - 10 | 1422 | 85 | ||
| BC020400.1 | - | 9 - 10 | 1392 | 85 | ||
| BC031700.1 | - | 9 - 10 | 1377 | 5007886 | 85 | |
| X62154.1 | - | 8 - 9 | 1321 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mcm3
Genomic Map