Assay Details
| Gene Symbol: Scn7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009135.2 | NP_033161.2 | 12 - 13 | 1841 | 127 | |
| GenBank mRNA | L36179.1 | - | 12 - 13 | 1877 | 127 | |
More Information
Set Membership: |
|

Scn7a
Genomic Map