Assay Details
| Gene Symbol: Ace | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001281819.1 | NP_001268748.1 | 15 - 16 | 2354 | 133 | |
| NM_009598.2 | NP_033728.1 | 4 - 5 | 577 | 133 | ||
| NM_207624.5 | NP_997507.1 | 15 - 16 | 2354 | 133 | ||
| GenBank mRNA | AK154632.1 | - | 15 - 16 | 2353 | 133 | |
| AK161020.1 | - | 15 - 16 | 2350 | 133 | ||
| AK164780.1 | - | 2 - 3 | 222 | 133 | ||
| BC034367.1 | - | 11 - 12 | 1616 | 133 | ||
| BC040404.1 | - | 15 - 16 | 2343 | 3965649 | 133 | |
| BC083109.1 | - | 15 - 16 | 2132 | 133 | ||
| BC110362.1 | - | 15 - 16 | 2340 | 5343978 | 133 | |
| J04946.1 | - | 9 - 10 | 1321 | 133 | ||
| J04947.1 | - | 1321 | 133 | |||
| M55333.1 | - | 4 - 5 | 577 | 133 | ||
More Information
Set Membership: |
|

Ace
Genomic Map