Assay Details
| Gene Symbol: Apbb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001201413.1 | NP_001188342.1 | 15 - 16 | 2501 | 62 | |
| NM_001201414.1 | NP_001188343.1 | 14 - 15 | 2504 | 62 | ||
| NM_001201415.1 | NP_001188344.1 | 14 - 15 | 2438 | 62 | ||
| NM_001201416.1 | NP_001188345.1 | 5 - 6 | 604 | 62 | ||
| NM_009686.2 | NP_033816.1 | 16 - 17 | 2507 | 62 | ||
| GenBank mRNA | AF133665.1 | - | 1 - 2 | 79 | 62 | |
| AK004792.1 | - | 16 - 17 | 2361 | 62 | ||
| AK153260.1 | - | 14 - 15 | 2470 | 62 | ||
| AK167649.1 | - | 15 - 16 | 2366 | 62 | ||
| AK169959.1 | - | 14 - 15 | 2376 | 62 | ||
| AK172405.1 | - | 5 - 6 | 608 | 62 | ||
| BC042797.1 | - | 6 - 7 | 646 | 62 | ||
| BC076587.1 | - | 14 - 15 | 2284 | 30550087 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Apbb2
Genomic Map