Assay Details
| Gene Symbol: Aqp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009700.2 | NP_033830.2 | 2 - 3 | 496 | 56 | |
| GenBank mRNA | AF469168.1 | - | 2 - 3 | 570 | 56 | |
| AF469169.1 | - | 2 - 3 | 752 | 56 | ||
| AK039777.1 | - | 2 - 3 | 499 | 56 | ||
| AK044074.1 | - | 2 - 3 | 435 | 56 | ||
| AK045357.1 | - | 1 - 2 | 147 | 56 | ||
| AY484964.1 | - | 2 - 3 | 502 | 56 | ||
| AY484965.1 | - | 3 - 4 | 686 | 56 | ||
| AY484966.1 | - | 3 - 4 | 632 | 56 | ||
| AY728043.1 | - | 2 - 3 | 516 | 56 | ||
| AY728044.1 | - | 3 - 4 | 700 | 56 | ||
| AY728045.1 | - | 2 - 3 | 628 | 56 | ||
| AY803942.1 | - | 3 - 4 | 607 | 56 | ||
| AY803943.1 | - | 4 - 5 | 737 | 56 | ||
| AY803944.1 | - | 5 - 6 | 921 | 56 | ||
| BB628835.1 | - | 2 - 3 | 497 | 56 | ||
| BC024526.1 | - | 2 - 3 | 442 | 4985265 | 56 | |
| CT010362.1 | - | 1 - 2 | 443 | 56 | ||
| U33012.1 | - | 1 - 2 | 458 | 56 | ||
| U48397.1 | - | 1 - 2 | 578 | 56 | ||
| U48398.1 | - | 1 - 2 | 449 | 56 | ||
| U48399.1 | - | 3 - 4 | 560 | 56 | ||
| U88623.1 | - | 2 - 3 | 497 | 56 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Aqp4
Genomic Map