Assay Details
| Gene Symbol: Arsa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009713.4 | NP_033843.2 | 4 - 5 | 1886 | 68 | |
| GenBank mRNA | AK004540.1 | - | 3 - 4 | 1105 | 68 | |
| AK132501.1 | - | 3 - 4 | 1117 | 68 | ||
| AK165921.1 | - | 3 - 4 | 1071 | 68 | ||
| BC011284.1 | - | 3 - 4 | 1068 | 3669438 | 68 | |
| BC098075.1 | - | 4 - 5 | 1898 | 30734943 | 68 | |
| X73230.1 | - | 3 - 4 | 1319 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arsa
Genomic Map