Assay Details
| Gene Symbol: Casp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080126.1 | NP_001073595.1 | 6 - 7 | 892 | 69 | |
| NM_001277926.1 | NP_001264855.1 | 7 - 8 | 1008 | 69 | ||
| NM_009812.2 | NP_033942.1 | 6 - 7 | 1065 | 69 | ||
| GenBank mRNA | AF067834.1 | - | 6 - 7 | 945 | 69 | |
| AJ000641.1 | - | 5 - 6 | 511 | 69 | ||
| AJ007749.1 | - | 6 - 7 | 718 | 69 | ||
| AK146758.1 | - | 6 - 7 | 1065 | 69 | ||
| AK153340.1 | - | 6 - 7 | 774 | 69 | ||
| AK156135.1 | - | 6 - 7 | 781 | 69 | ||
| AK168812.1 | - | 6 - 7 | 1065 | 69 | ||
| BC006737.1 | - | 6 - 7 | 1007 | 3964119 | 69 | |
| BC049955.1 | - | 6 - 7 | 899 | 4208824 | 69 | |
| CT010166.1 | - | 5 - 6 | 672 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Casp8
Genomic Map